N-(3-Acetylphenyl)-2-cyanoacetamide

Supplier: Matrix Scientific

039641-500MG
101847-892EA 109.25 USD
101847-892
N-(3-Acetylphenyl)-2-cyanoacetamide
N-(3-Acetylphenyl)-2-cyanoacetamide

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MDL Number: MFCD03362300
CAS Number: 540532-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR