4-Cyano-5-[(cyanoacetyl)amino]-N,N-diethyl-3-methylthiophene-2-carboxamide

Supplier: Matrix Scientific

039640-500MG
101847-890EA 387.69 USD
101847-890
4-Cyano-5-[(cyanoacetyl)amino]-N,N-diethyl-3-methylthiophene-2-carboxamide
4-Cyano-5-[(cyanoacetyl)amino]-N,N-diethyl-3-methylthiophene-2-carboxamide

Hazard: Irritant

Formula: C₁₄H₁₆N₄O₂S
MDL Number: MFCD01358310
CAS Number: 581780-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR