2-Cyano-N-[3-(morpholin-4-ylcarbonyl)-5,6,7,8-tetrahydro-4H-cyclohepta[b]thien-2-yl]acetamide

Supplier: Matrix Scientific

039639-500MG
101847-888EA 289.8 USD
101847-888
2-Cyano-N-[3-(morpholin-4-ylcarbonyl)-5,6,7,8-tetrahydro-4H-cyclohepta[b]thien-2-yl]acetamide
2-Cyano-N-[3-(morpholin-4-ylcarbonyl)-5,6,7,8-tetrahydro-4H-cyclohepta[b]thien-2-yl]acetamide

Hazard: Irritant

Formula: C₁₇H₂₁N₃O₃S
MDL Number: MFCD03945359
CAS Number: 588715-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR