3-[4-(Diphenylmethyl)piperazin-1-yl]-3-oxopropanenitrile

Supplier: Matrix Scientific

039638-500MG
101847-886EA 215.38 USD
101847-886
3-[4-(Diphenylmethyl)piperazin-1-yl]-3-oxopropanenitrile
3-[4-(Diphenylmethyl)piperazin-1-yl]-3-oxopropanenitrile

Hazard: Irritant

Formula: C₂₀H₂₁N₃O
MDL Number: MFCD01341785
CAS Number: 313386-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR