Methyl-2-[(cyanoacetyl)amino]-5-[(diethylamino)carbonyl]-4-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

039637-500MG
101847-884EA 289.8 USD
101847-884
Methyl-2-[(cyanoacetyl)amino]-5-[(diethylamino)carbonyl]-4-methylthiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-5-[(diethylamino)carbonyl]-4-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₉N₃O₄S
MDL Number: MFCD03757168
CAS Number: 514218-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR