2-Cyano-N-(3-cyano-4,5,6,7-tetrahydro-1-benzothien-2-yl)acetamide

Supplier: Matrix Scientific

039634-500MG
101847-878EA 217.35 USD
101847-878
2-Cyano-N-(3-cyano-4,5,6,7-tetrahydro-1-benzothien-2-yl)acetamide
2-Cyano-N-(3-cyano-4,5,6,7-tetrahydro-1-benzothien-2-yl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₁N₃OS
MDL Number: MFCD00835750
CAS Number: 73227-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR