Ethyl-2-[(cyanoacetyl)amino]-5-[(dimethylamino)-carbonyl]-4-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

039633-500MG
101847-876EA 387.69 USD
101847-876
Ethyl-2-[(cyanoacetyl)amino]-5-[(dimethylamino)-carbonyl]-4-methylthiophene-3-carboxylate
Ethyl-2-[(cyanoacetyl)amino]-5-[(dimethylamino)-carbonyl]-4-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₇N₃O₄S
MDL Number: MFCD03389930
CAS Number: 545372-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR