Diethyl-5-[(cyanoacetyl)amino]-3-methylthiophene-2,4-dicarboxylate

Supplier: Matrix Scientific

039632-500MG
101847-874EA 289.8 USD
101847-874
Diethyl-5-[(cyanoacetyl)amino]-3-methylthiophene-2,4-dicarboxylate
Diethyl-5-[(cyanoacetyl)amino]-3-methylthiophene-2,4-dicarboxylate

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₅S
MDL Number: MFCD03757173
CAS Number: 514218-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR