Methyl-2-[(cyanoacetyl)amino]-5-isopropylthiophene-3-carboxylate

Supplier: Matrix Scientific

039626-500MG
101847-862EA 289.8 USD
101847-862
Methyl-2-[(cyanoacetyl)amino]-5-isopropylthiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-5-isopropylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃S
MDL Number: MFCD03759393
CAS Number: 515861-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR