2-Cyano-N-(2-ethoxyphenyl)acetamide

Supplier: Matrix Scientific

039621-500MG
101847-852EA 146.15 USD
101847-852
2-Cyano-N-(2-ethoxyphenyl)acetamide
2-Cyano-N-(2-ethoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD03150623
CAS Number: 380427-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR