N-(2-Chlorophenyl)-2-cyanoacetamide

Supplier: Matrix Scientific

039620-500MG
101847-850EA 109.25 USD
101847-850
N-(2-Chlorophenyl)-2-cyanoacetamide
N-(2-Chlorophenyl)-2-cyanoacetamide

Hazard: Irritant

Formula: C₉H₇ClN₂O
MDL Number: MFCD00463630
CAS Number: 50982-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR