Methyl-5-benzyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Supplier: Matrix Scientific

039618-500MG
101847-846EA 387.69 USD
101847-846
Methyl-5-benzyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate
Methyl-5-benzyl-2-[(cyanoacetyl)amino]thiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₄N₂O₃S
MDL Number: MFCD03759462
CAS Number: 515872-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR