2-Cyano-N-(2,5-dimethoxyphenyl)acetamide

Supplier: Matrix Scientific

039612-500MG
101847-834EA 146.15 USD
101847-834
2-Cyano-N-(2,5-dimethoxyphenyl)acetamide
2-Cyano-N-(2,5-dimethoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃
MDL Number: MFCD03382982
CAS Number: 113604-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR