2-Cyano-N-(2,4-dichlorophenyl)acetamide

Supplier: Matrix Scientific

039611-500MG
101847-832EA 146.15 USD
101847-832
2-Cyano-N-(2,4-dichlorophenyl)acetamide
2-Cyano-N-(2,4-dichlorophenyl)acetamide

Hazard: Irritant

Formula: C₉H₆Cl₂N₂O
MDL Number: MFCD00170778
CAS Number: 17722-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR