Methyl-2-[(cyanoacetyl)amino]-5-[(dimethylamino)carbonyl]-4-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

039608-500MG
101847-826EA 146.15 USD
101847-826
Methyl-2-[(cyanoacetyl)amino]-5-[(dimethylamino)carbonyl]-4-methylthiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-5-[(dimethylamino)carbonyl]-4-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₄S
MDL Number: MFCD03394772
CAS Number: 546090-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR