Methyl-5-(aminocarbonyl)-2-[(cyanoacetyl)amino]-4-methylthiophene-3-carboxylate

Supplier: Matrix Scientific

039607-500MG
101847-824EA 146.15 USD
101847-824
Methyl-5-(aminocarbonyl)-2-[(cyanoacetyl)amino]-4-methylthiophene-3-carboxylate
Methyl-5-(aminocarbonyl)-2-[(cyanoacetyl)amino]-4-methylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁N₃O₄S
MDL Number: MFCD03759373
CAS Number: 515860-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR