N-(6-tert-Butyl-3-cyano-4,5,6,7-tetrahydro-1-benzothien-2-yl)-2-cyanoacetamide

Supplier: Matrix Scientific

039606-500MG
101847-822EA 109.25 USD
101847-822
N-(6-tert-Butyl-3-cyano-4,5,6,7-tetrahydro-1-benzothien-2-yl)-2-cyanoacetamide
N-(6-tert-Butyl-3-cyano-4,5,6,7-tetrahydro-1-benzothien-2-yl)-2-cyanoacetamide

Hazard: Irritant

Formula: C₁₆H₁₉N₃OS
MDL Number: MFCD03385360
CAS Number: 560072-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR