Methyl-2-[(cyanoacetyl)amino]-5-methylthiophene-3-carboxylate
Supplier: Matrix Scientific
039605-500MG
101847-820EA
146.15
USD
101847-820
Methyl-2-[(cyanoacetyl)amino]-5-methylthiophene-3-carboxylate
Methyl-2-[(cyanoacetyl)amino]-5-methylthiophene-3-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₃S |
MDL Number:
MFCD03759397 CAS Number: 515861-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...