N-(2-Chloro-5-nitrophenyl)-2-cyanoacetamide

Supplier: Matrix Scientific

039604-500MG
101847-818EA 109.25 USD
101847-818
N-(2-Chloro-5-nitrophenyl)-2-cyanoacetamide
N-(2-Chloro-5-nitrophenyl)-2-cyanoacetamide

Hazard: Irritant

Formula: C₉H₆ClN₃O₃
MDL Number: MFCD03382108
CAS Number: 544669-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR