2-Cyano-N-[1-(4-ethylphenyl)ethyl]acetamide

Supplier: Matrix Scientific

039601-500MG
101847-812EA 109.25 USD
101847-812
2-Cyano-N-[1-(4-ethylphenyl)ethyl]acetamide
2-Cyano-N-[1-(4-ethylphenyl)ethyl]acetamide

Hazard: Irritant

Formula: C₁₃H₁₆N₂O
MDL Number: MFCD02177236
CAS Number: 444907-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR