2-Cyano-N-(4-methylbenzyl)acetamide

Supplier: Matrix Scientific

039600-500MG
101847-810EA 146.15 USD
101847-810
2-Cyano-N-(4-methylbenzyl)acetamide
2-Cyano-N-(4-methylbenzyl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD01349939
CAS Number: 64488-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR