2-Cyano-N-[1-(4-methylphenyl)ethyl]acetamide

Supplier: Matrix Scientific

039595-500MG
101847-800EA 146.15 USD
101847-800
2-Cyano-N-[1-(4-methylphenyl)ethyl]acetamide
2-Cyano-N-[1-(4-methylphenyl)ethyl]acetamide

Hazard: Irritant

Formula: C₁₂H₁₄N₂O
MDL Number: MFCD02177127
CAS Number: 444907-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR