2-Cyano-N-cyclopropylacetamide

Supplier: Matrix Scientific

039593-500MG
101847-796EA 230.77 USD
101847-796
2-Cyano-N-cyclopropylacetamide
2-Cyano-N-cyclopropylacetamide

Hazard: Irritant

Formula: C₆H₈N₂O
MDL Number: MFCD02176352
CAS Number: 15029-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR