2-Cyano-N-(3-pyridinylmethyl)acetamide

Supplier: Matrix Scientific

039592-500MG
101847-794EA 146.15 USD
101847-794
2-Cyano-N-(3-pyridinylmethyl)acetamide
2-Cyano-N-(3-pyridinylmethyl)acetamide

Hazard: Irritant

Formula: C₉H₉N₃O
MW: 175.19 g/mol
MDL Number: MFCD01351849
CAS Number: 283153-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR