3-[4-(3,4-Dichlorophenyl)piperazin-1-yl]-3-oxopropanenitrile
Supplier: Matrix Scientific
039591-500MG
101847-792EA
146.15
USD
101847-792
3-[4-(3,4-Dichlorophenyl)piperazin-1-yl]-3-oxopropanenitrile
3-[4-(3,4-Dichlorophenyl)piperazin-1-yl]-3-oxopropanenitrile
Hazard: Irritant
Formula:
C₁₃H₁₃Cl₂N₃O |
MDL Number:
MFCD01993790 CAS Number: 350999-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...