2-Cyano-N-propylacetamide
039589-500MG
101847-788EA
579.58
USD
101847-788
2-Cyano-N-propylacetamide
2-Cyano-N-propylacetamide
Hazard: Irritant
Formula:
C₆H₁₀N₂O |
MDL Number:
MFCD02176952 CAS Number: 52493-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...