3-[4-(4-Methoxyphenyl)piperazin-1-yl]-3-oxopropanenitrile

Supplier: Matrix Scientific

039587-500MG
101847-784EA 146.15 USD
101847-784
3-[4-(4-Methoxyphenyl)piperazin-1-yl]-3-oxopropanenitrile
3-[4-(4-Methoxyphenyl)piperazin-1-yl]-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₄H₁₇N₃O₂
MDL Number: MFCD02176531
CAS Number: 524926-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR