Methyl-2-mercapto-4-(methoxymethyl)-1,3-thiazole-5-carboxylate

Supplier: Matrix Scientific

039585-500MG
101847-780EA 387.69 USD
101847-780
Methyl-2-mercapto-4-(methoxymethyl)-1,3-thiazole-5-carboxylate
Methyl-2-mercapto-4-(methoxymethyl)-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₉NO₃S₂
MDL Number: MFCD12028045
CAS Number: 1142201-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR