Methyl-2-amino-4-(methoxymethyl)-1,3-thiazole-5-carboxylate

Supplier: Matrix Scientific

039584-500MG
101847-778EA 289.8 USD
101847-778
Methyl-2-amino-4-(methoxymethyl)-1,3-thiazole-5-carboxylate
Methyl-2-amino-4-(methoxymethyl)-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₃S
MW: 202.23 g/mol
MDL Number: MFCD12028044
CAS Number: 733754-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR