(5E)-5-[(2-Aminopyridin-3-yl)methylene]-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039580-500MG
101847-770EA 144.9 USD
101847-770
(5E)-5-[(2-Aminopyridin-3-yl)methylene]-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-[(2-Aminopyridin-3-yl)methylene]-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₅H₁₂N₄OS
MDL Number: MFCD12028040
CAS Number: 1142201-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR