(5E)-2-Mercapto-5-[(6-methylpyridin-2-yl)methylene]-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Supplier: Matrix Scientific
039578-500MG
101847-766EA
193.85
USD
101847-766
(5E)-2-Mercapto-5-[(6-methylpyridin-2-yl)methylene]-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-2-Mercapto-5-[(6-methylpyridin-2-yl)methylene]-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Hazard: Irritant
Formula:
C₁₆H₁₃N₃OS |
MDL Number:
MFCD12028038 CAS Number: 1142201-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...