(5E)-5-(1H-Indol-5-ylmethylene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Supplier: Matrix Scientific
039575-500MG
101847-760EA
193.85
USD
101847-760
(5E)-5-(1H-Indol-5-ylmethylene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(1H-Indol-5-ylmethylene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Hazard: Irritant
Formula:
C₁₈H₁₃N₃OS |
MDL Number:
MFCD12028035 CAS Number: 1142201-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...