(5E)-5-(3,4-Dihydroxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039573-500MG
101847-756EA 193.85 USD
101847-756
(5E)-5-(3,4-Dihydroxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(3,4-Dihydroxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₆H₁₂N₂O₃S
MDL Number: MFCD12028033
CAS Number: 897340-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR