(5E)-2-Mercapto-5-[(3-methyl-2-thienyl)methylene]-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039571-500MG
101847-752EA 193.85 USD
101847-752
(5E)-2-Mercapto-5-[(3-methyl-2-thienyl)methylene]-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-2-Mercapto-5-[(3-methyl-2-thienyl)methylene]-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₅H₁₂N₂OS₂
MDL Number: MFCD12028031
CAS Number: 364614-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR