(5E)-2-Mercapto-5-[(5-methyl-2-furyl)methylene]-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039569-500MG
101847-748EA 193.85 USD
101847-748
(5E)-2-Mercapto-5-[(5-methyl-2-furyl)methylene]-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-2-Mercapto-5-[(5-methyl-2-furyl)methylene]-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₅H₁₂N₂O₂S
MDL Number: MFCD12028029
CAS Number: 341943-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR