(5E)-5-(2,1,3-Benzothiadiazol-4-ylmethylene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039568-500MG
101847-746EA 193.85 USD
101847-746
(5E)-5-(2,1,3-Benzothiadiazol-4-ylmethylene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(2,1,3-Benzothiadiazol-4-ylmethylene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₆H₁₀N₄OS₂
MDL Number: MFCD12028028
CAS Number: 1142201-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR