(5E)-5-(4-Bromobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Supplier: Matrix Scientific
039563-500MG
101847-736EA
869.23
USD
101847-736
(5E)-5-(4-Bromobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(4-Bromobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Hazard: Irritant
Formula:
C₁₆H₁₁BrN₂OS |
MDL Number:
MFCD12028023 CAS Number: 423741-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...