(5E)-5-(3-Bromobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039562-500MG
101847-734EA 144.9 USD
101847-734
(5E)-5-(3-Bromobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(3-Bromobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₆H₁₁BrN₂OS
MDL Number: MFCD12028022
CAS Number: 301343-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR