(5E)-2-Mercapto-3-phenyl-5-(2,4,5-trimethoxy-benzylidene)-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039559-500MG
101847-728EA 193.85 USD
101847-728
(5E)-2-Mercapto-3-phenyl-5-(2,4,5-trimethoxy-benzylidene)-3,5-dihydro-4H-imidazol-4-one
(5E)-2-Mercapto-3-phenyl-5-(2,4,5-trimethoxy-benzylidene)-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₉H₁₈N₂O₄S
MDL Number: MFCD12028019
CAS Number: 94451-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR