(5E)-2-Mercapto-5-(4-methylbenzylidene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039558-500MG
101847-726EA 193.85 USD
101847-726
(5E)-2-Mercapto-5-(4-methylbenzylidene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-2-Mercapto-5-(4-methylbenzylidene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₇H₁₄N₂OS
MDL Number: MFCD08741783
CAS Number: 13109-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR