(5E)-2-Mercapto-3-phenyl-5-(pyridin-3-ylmethylene)-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039553-500MG
101847-716EA 193.85 USD
101847-716
(5E)-2-Mercapto-3-phenyl-5-(pyridin-3-ylmethylene)-3,5-dihydro-4H-imidazol-4-one
(5E)-2-Mercapto-3-phenyl-5-(pyridin-3-ylmethylene)-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₅H₁₁N₃OS
MDL Number: MFCD12028014
CAS Number: 145693-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR