(5E)-2-Mercapto-3-phenyl-5-(3-thienylmethylene)-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039552-500MG
101847-714EA 144.9 USD
101847-714
(5E)-2-Mercapto-3-phenyl-5-(3-thienylmethylene)-3,5-dihydro-4H-imidazol-4-one
(5E)-2-Mercapto-3-phenyl-5-(3-thienylmethylene)-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₄H₁₀N₂OS₂
MDL Number: MFCD12028013
CAS Number: 890273-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR