(5E)-2-Mercapto-5-(4-nitrobenzylidene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039551-500MG
101847-712EA 144.9 USD
101847-712
(5E)-2-Mercapto-5-(4-nitrobenzylidene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-2-Mercapto-5-(4-nitrobenzylidene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₆H₁₁N₃O₃S
MDL Number: MFCD12028012
CAS Number: 53514-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR