(5E)-2-Mercapto-5-(1-naphthylmethylene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Supplier: Matrix Scientific
039550-500MG
101847-710EA
193.85
USD
101847-710
(5E)-2-Mercapto-5-(1-naphthylmethylene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-2-Mercapto-5-(1-naphthylmethylene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Hazard: Irritant
Formula:
C₂₀H₁₄N₂OS |
MDL Number:
MFCD12028011 CAS Number: 61442-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...