(5E)-5-(2-Furylmethylene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Supplier: Matrix Scientific
039548-500MG
101847-706EA
193.85
USD
101847-706
(5E)-5-(2-Furylmethylene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(2-Furylmethylene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Hazard: Irritant
Formula:
C₁₄H₁₀N₂O₂S |
MDL Number:
MFCD12028009 CAS Number: 87736-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...