(5E)-5-(4-Ethoxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Supplier: Matrix Scientific
039545-500MG
101847-700EA
193.85
USD
101847-700
(5E)-5-(4-Ethoxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(4-Ethoxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Hazard: Irritant
Formula:
C₁₈H₁₆N₂O₂S |
MDL Number:
MFCD12028008 CAS Number: 894163-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...