(5E)-5-[4-(Dimethylamino)benzylidene]-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039540-500MG
101847-690EA 144.9 USD
101847-690
(5E)-5-[4-(Dimethylamino)benzylidene]-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-[4-(Dimethylamino)benzylidene]-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₈H₁₇N₃OS
MDL Number: MFCD12028004
CAS Number: 301343-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR