(5E)-5-(2,5-Dimethoxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Supplier: Matrix Scientific
039538-500MG
101847-686EA
193.85
USD
101847-686
(5E)-5-(2,5-Dimethoxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(2,5-Dimethoxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
Hazard: Irritant
Formula:
C₁₈H₁₆N₂O₃S |
MDL Number:
MFCD12028003 CAS Number: 15985-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...