(5E)-5-(2,4-Dimethoxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039537-500MG
101847-684EA 193.85 USD
101847-684
(5E)-5-(2,4-Dimethoxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(2,4-Dimethoxybenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₈H₁₆N₂O₃S
MDL Number: MFCD12028002
CAS Number: 402608-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR