(5E)-5-(2,4-Dichlorobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039534-500MG
101847-678EA 193.85 USD
101847-678
(5E)-5-(2,4-Dichlorobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(2,4-Dichlorobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₆H₁₀Cl₂N₂OS
MDL Number: MFCD12027999
CAS Number: 444061-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR